Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jmjd7
Entrez Gene ID
496732
Gene Name
jumonji domain containing 7
Gene Aliases
-
Chromosome Location
Chr.8: 84704579 - 84710994 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011279.1 NP_001011279.1 2 - 3 237 65
GenBank mRNA BC087993.1 - 2 - 3 237 65

View Full Product Details