Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ela3b
Entrez Gene ID
496902
Gene Name
elastase 3B, pancreatic
Gene Aliases
-
Chromosome Location
Chr.7: 116188830 - 116190085 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011421.1 NP_001011421.1 - 146 110
GenBank mRNA BC087729.1 - - 136 110
BC088036.1 - - 146 110

View Full Product Details