Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc50a1
Entrez Gene ID
548582
Gene Name
solute carrier family 50 (sugar efflux transporter), member 1
Gene Aliases
rag1ap1
Chromosome Location
Chr.Un|NW_016685101: 1836 - 5241 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015865.1 NP_001015865.1 1 - 2 301 68
GenBank mRNA BC090379.1 - 1 - 2 301 68

View Full Product Details