Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ocln
Entrez Gene ID
548702
Gene Name
occludin
Gene Aliases
ocln-a, ocln-b, tpmt
Chromosome Location
Chr.1: 167342796 - 167364715 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015948.1 NP_001015948.1 8 - 9 1596 92
GenBank mRNA BC123964.1 - 8 - 9 1589 92
CR761445.2 - 8 - 9 1596 92

View Full Product Details