Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ucma
Entrez Gene ID
549239
Gene Name
upper zone of growth plate and cartilage matrix associated
Gene Aliases
grp
Chromosome Location
Chr.3: 72254962 - 72262282 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016485.2 NP_001016485.1 1 - 2 54 86
GenBank mRNA BC135882.1 - 1 - 2 68 86
CR760848.2 - 1 - 2 54 86

View Full Product Details