Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbm39
Entrez Gene ID
549542
Gene Name
RNA binding motif protein 39
Gene Aliases
caper, caperalpha, cc1.3, hcc1, rnpc2
Chromosome Location
Chr.10: 34295850 - 34311608 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016788.2 NP_001016788.1 11 - 12 1407 100
XM_012971783.2 XP_012827237.1 11 - 12 1444 100
XM_012971784.2 XP_012827238.1 11 - 12 1441 100
XM_012971785.2 XP_012827239.1 12 - 13 1609 100
XM_012971786.2 XP_012827240.1 11 - 12 1426 100
XM_012971788.2 XP_012827242.1 12 - 13 1557 100
XM_012971789.2 XP_012827243.1 13 - 14 1670 100
XM_012971790.2 XP_012827244.1 12 - 13 1516 100
XM_012971792.2 XP_012827246.1 12 - 13 1541 100
XM_018097748.1 XP_017953237.1 11 - 12 2833 100
XM_018097749.1 XP_017953238.1 12 - 13 2160 100
XM_018097750.1 XP_017953239.1 12 - 13 2108 100
XM_018097751.1 XP_017953240.1 12 - 13 2067 100
XM_018097752.1 XP_017953241.1 13 - 14 1712 100
XM_018097753.1 XP_017953242.1 11 - 12 1444 100
XM_018097754.1 XP_017953243.1 12 - 13 1517 100
XM_018097755.1 XP_017953244.1 11 - 12 1994 100
XM_018097756.1 XP_017953245.1 12 - 13 1610 100
XM_018097757.1 XP_017953246.1 12 - 13 1558 100
XM_018097758.1 XP_017953247.1 11 - 12 2819 100
XM_018097759.1 XP_017953248.1 12 - 13 2858 100
GenBank mRNA BC093451.1 - 11 - 12 1407 100
CR855673.2 - 11 - 12 1310 100

View Full Product Details