Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rbm39
Entrez Gene ID
549542
Gene Name
RNA binding motif protein 39
Gene Aliases
caper, caperalpha, cc1.3, hcc1, rnpc2
Chromosome Location
Chr.10: 34295850 - 34311608 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016788.2 NP_001016788.1 4 - 5 606 91
XM_012971783.2 XP_012827237.1 4 - 5 628 91
XM_012971784.2 XP_012827238.1 4 - 5 628 91
XM_012971785.2 XP_012827239.1 5 - 6 793 91
XM_012971786.2 XP_012827240.1 4 - 5 628 91
XM_012971788.2 XP_012827242.1 5 - 6 741 91
XM_012971789.2 XP_012827243.1 6 - 7 854 91
XM_012971790.2 XP_012827244.1 5 - 6 700 91
XM_012971792.2 XP_012827246.1 5 - 6 740 91
XM_018097748.1 XP_017953237.1 4 - 5 2017 91
XM_018097749.1 XP_017953238.1 5 - 6 1344 91
XM_018097750.1 XP_017953239.1 5 - 6 1292 91
XM_018097751.1 XP_017953240.1 5 - 6 1251 91
XM_018097752.1 XP_017953241.1 6 - 7 896 91
XM_018097753.1 XP_017953242.1 4 - 5 628 91
XM_018097754.1 XP_017953243.1 5 - 6 701 91
XM_018097755.1 XP_017953244.1 4 - 5 1178 91
XM_018097756.1 XP_017953245.1 5 - 6 794 91
XM_018097757.1 XP_017953246.1 5 - 6 742 91
XM_018097758.1 XP_017953247.1 4 - 5 2018 91
GenBank mRNA BC093451.1 - 4 - 5 606 91
CR855673.2 - 4 - 5 509 91

View Full Product Details