Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
azin2
Entrez Gene ID
548747
Gene Name
antizyme inhibitor 2
Gene Aliases
adc, azi2, odc-p, odc1l, odc2, xodc2
Chromosome Location
Chr.2: 86263910 - 86270139 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015993.3 NP_001015993.2 8 - 9 1194 110
GenBank mRNA CR848307.2 - 8 - 9 1133 110
DT426956.1 - 4 - 5 620 110

View Full Product Details