Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vps25
Entrez Gene ID
548832
Gene Name
vacuolar protein sorting 25 homolog
Gene Aliases
derp9, eap20, fap20
Chromosome Location
Chr.10: 11422348 - 11432652 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016078.2 NP_001016078.1 2 - 3 203 72
GenBank mRNA BC121525.1 - 2 - 3 205 72
CR762222.2 - 2 - 3 203 72

View Full Product Details