Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tor1b
Entrez Gene ID
548991
Gene Name
torsin family 1, member B (torsin B)
Gene Aliases
tor1a
Chromosome Location
Chr.8: 4485832 - 4491750 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016237.2 NP_001016237.1 3 - 4 687 88
GenBank mRNA BC161275.1 - 3 - 4 622 88
BC170808.1 - 3 - 4 672 88
BC171143.1 - 3 - 4 672 88
CR926267.2 - 3 - 4 687 88

View Full Product Details