Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kti12
Entrez Gene ID
549045
Gene Name
KTI12 chromatin associated homolog
Gene Aliases
sbbi81, tot4
Chromosome Location
Chr.3: 81044638 - 81072227 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016291.2 NP_001016291.1 4 - 5 296 89
XM_012966570.1 XP_012822024.1 4 - 5 490 89
XM_018092083.1 XP_017947572.1 4 - 5 329 89
GenBank mRNA BC170877.1 - 3 - 4 276 89
BC170879.1 - 3 - 4 276 89
CR761570.2 - 4 - 5 296 89

View Full Product Details