Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cenpa
Entrez Gene ID
549339
Gene Name
centromere protein A
Gene Aliases
cenp-a, cenpx
Chromosome Location
Chr.8: 54189039 - 54198980 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016585.2 NP_001016585.1 1 - 2 87 73
XM_012968427.1 XP_012823881.1 2 - 3 430 73
XM_012968428.2 XP_012823882.1 2 - 3 178 73
XM_012968429.1 XP_012823883.1 2 - 3 454 73
XM_012968430.2 XP_012823884.1 2 - 3 147 73
GenBank mRNA CR760620.2 - 1 - 2 87 73

View Full Product Details