Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zc4h2
Entrez Gene ID
549429
Gene Name
zinc finger, C4H2 domain containing
Gene Aliases
hca127
Chromosome Location
Chr.8: 25054112 - 25068654 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016675.2 NP_001016675.1 2 - 3 269 79
GenBank mRNA BC170701.1 - 2 - 3 258 79
BC171113.1 - 2 - 3 258 79
CR760233.2 - 2 - 3 269 79

View Full Product Details