Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glrx3
Entrez Gene ID
549963
Gene Name
glutaredoxin 3
Gene Aliases
picot, txnl2
Chromosome Location
Chr.7: 13875686 - 13903069 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017209.2 NP_001017209.1 5 - 6 623 84
GenBank mRNA BC155988.1 - 5 - 6 605 84
CR760465.2 - 5 - 6 623 84

View Full Product Details