Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klhl8
Entrez Gene ID
550043
Gene Name
kelch like family member 8
Gene Aliases
-
Chromosome Location
Chr.1: 93702356 - 93749230 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017289.2 NP_001017289.1 4 - 5 1171 61
XM_018093771.1 XP_017949260.1 5 - 6 2647 61
XM_018093773.1 XP_017949262.1 4 - 5 960 61
GenBank mRNA BC167576.1 - 5 - 6 1214 61
CR855674.2 - 4 - 5 1171 61

View Full Product Details