Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spsb3
Entrez Gene ID
550104
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
-
Chromosome Location
Chr.9: 15565897 - 15575193 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017350.2 NP_001017350.1 4 - 5 626 77
XM_012970482.2 XP_012825936.1 5 - 6 701 77
XM_012970483.2 XP_012825937.1 5 - 6 804 77
XM_012970484.2 XP_012825938.1 5 - 6 698 77
XM_018097133.1 XP_017952622.1 5 - 6 691 77
GenBank mRNA BC160472.1 - 5 - 6 674 77
CR848600.2 - 4 - 5 626 77

View Full Product Details