Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
thrap3
Entrez Gene ID
394509
Gene Name
thyroid hormone receptor associated protein 3
Gene Aliases
trap150
Chromosome Location
Chr.2: 55434378 - 55472130 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037250.1 NP_001032327.1 6 - 7 1841 74
XM_012956627.2 XP_012812081.1 7 - 8 1967 74
XM_012956628.2 XP_012812082.1 6 - 7 1908 74
XM_012956629.2 XP_012812083.1 7 - 8 1964 74
XM_012956630.1 XP_012812084.1 6 - 7 1921 74
XM_012956631.1 XP_012812085.1 6 - 7 1921 74
XM_012956632.1 XP_012812086.1 6 - 7 1921 74
XM_012956633.1 XP_012812087.1 6 - 7 1921 74
XM_012956634.1 XP_012812088.1 6 - 7 1921 74
XM_018091133.1 XP_017946622.1 7 - 8 2038 74
XM_018091134.1 XP_017946623.1 6 - 7 1864 74
GenBank mRNA BC171045.1 - 6 - 7 1804 74
CR759994.2 - 6 - 7 1841 74

View Full Product Details