Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dgat2
Entrez Gene ID
395003
Gene Name
diacylglycerol O-acyltransferase 2
Gene Aliases
hmfn1045
Chromosome Location
Chr.2: 169512851 - 169520160 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204041.2 NP_989372.1 6 - 7 985 76
GenBank mRNA BC064191.1 - 6 - 7 982 76
CR855594.2 - 6 - 7 985 76

View Full Product Details