Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp1cc
Entrez Gene ID
407879
Gene Name
protein phosphatase 1, catalytic subunit, gamma isozyme
Gene Aliases
PP-1G, ppp1g
Chromosome Location
Chr.1: 154292568 - 154305260 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213670.2 NP_998835.1 2 - 3 371 93
GenBank mRNA BC067911.1 - 2 - 3 258 93
CR761289.1 - 2 - 3 371 93

View Full Product Details