Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem97
Entrez Gene ID
448515
Gene Name
transmembrane protein 97
Gene Aliases
-
Chromosome Location
Chr.2: 12563413 - 12568444 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005014.2 NP_001005014.1 1 - 2 192 83
GenBank mRNA BC076679.1 - 1 - 2 176 83
CR848476.1 - 1 - 2 192 83

View Full Product Details