Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arl16
Entrez Gene ID
733846
Gene Name
ADP ribosylation factor like GTPase 16
Gene Aliases
-
Chromosome Location
Chr.10: 19612085 - 19623712 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045595.1 NP_001039060.1 3 - 4 297 79
GenBank mRNA BC153678.1 - 3 - 4 336 79
CR848644.1 - 3 - 4 297 79

View Full Product Details