Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmed1
Entrez Gene ID
734138
Gene Name
transmembrane p24 trafficking protein 1
Gene Aliases
Xp24g1, Xp24gamma1
Chromosome Location
Chr.3: 127981365 - 127992048 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045798.1 NP_001039263.1 3 - 4 475 69
GenBank mRNA BC123004.1 - 3 - 4 467 69
CR942696.2 - 3 - 4 475 69

View Full Product Details