Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab40b
Entrez Gene ID
779634
Gene Name
RAB40B, member RAS oncogene family
Gene Aliases
rab40, rar, sec4l, xrab40
Chromosome Location
Chr.Un|NW_016683632: 72807 - 106259 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078720.1 NP_001072188.1 6 - 7 698 76
XM_018090185.1 XP_017945674.1 6 - 7 1235 76
GenBank mRNA BC118721.1 - 6 - 7 698 76
CT025300.2 - 6 - 7 700 76

View Full Product Details