Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
taf4
Entrez Gene ID
779738
Gene Name
TATA-box binding protein associated factor 4
Gene Aliases
taf2c, taf2c1, taf4a, tafii130, tafii135
Chromosome Location
Chr.10: 1088744 - 1118558 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078817.1 NP_001072285.1 8 - 9 2070 71
XM_012952548.2 XP_012808002.1 8 - 9 2639 71
GenBank mRNA BC121308.1 - 8 - 9 2070 71
CR761139.2 - 5 - 6 637 71

View Full Product Details