Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zbtb44
Entrez Gene ID
779914
Gene Name
zinc finger and BTB domain containing 44
Gene Aliases
btbd15, hspc063
Chromosome Location
Chr.7: 46804798 - 46844655 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078991.1 NP_001072459.1 3 - 4 1248 73
XM_012966742.2 XP_012822196.1 3 - 4 2218 73
GenBank mRNA BC121859.1 - 3 - 4 1248 73

View Full Product Details