Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ccdc69
Entrez Gene ID
780085
Gene Name
coiled-coil domain containing 69
Gene Aliases
ccdc69-a, ccdc69-b
Chromosome Location
Chr.3: 10969657 - 10991777 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079161.1 NP_001072629.1 8 - 9 810 74
GenBank mRNA BC123063.1 - 8 - 9 810 74
CT030062.2 - 8 - 9 864 74

View Full Product Details