Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sh2d4a
Entrez Gene ID
780161
Gene Name
SH2 domain containing 4A
Gene Aliases
sh2a
Chromosome Location
Chr.1: 77326575 - 77372443 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079236.2 NP_001072704.1 7 - 8 1222 97
XM_012965810.2 XP_012821264.1 8 - 9 1359 97
XM_012965811.2 XP_012821265.1 9 - 10 1462 97
XM_012965812.2 XP_012821266.1 8 - 9 1309 97
XM_012965813.2 XP_012821267.1 7 - 8 1281 97
GenBank mRNA BC124048.1 - 7 - 8 1223 97

View Full Product Details