Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a42
Entrez Gene ID
780169
Gene Name
solute carrier family 25 member 42
Gene Aliases
-
Chromosome Location
Chr.1: 102099446 - 102117751 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079244.1 NP_001072712.1 5 - 6 526 69
XM_012965828.2 XP_012821282.1 6 - 7 773 69
GenBank mRNA BC124065.1 - 5 - 6 526 69

View Full Product Details