Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
znrf3
Entrez Gene ID
780325
Gene Name
zinc and ring finger 3
Gene Aliases
-
Chromosome Location
Chr.1: 157585880 - 157671305 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079396.1 NP_001072864.1 5 - 6 988 93
XM_012966110.2 XP_012821564.1 4 - 5 4438 93
GenBank mRNA BC123917.1 - 5 - 6 988 93

View Full Product Details