Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
camlg
Entrez Gene ID
549354
Gene Name
calcium modulating ligand
Gene Aliases
-
Chromosome Location
Chr.3: 17125242 - 17131037 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016600.4 NP_001016600.2 1 - 2 281 99
GenBank mRNA BC136163.1 - 1 - 2 281 99
CR760559.2 - 1 - 2 189 99

View Full Product Details