Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
b3glct
Entrez Gene ID
780006
Gene Name
beta 3-glucosyltransferase
Gene Aliases
b3galtl
Chromosome Location
Chr.2: 150789562 - 150976728 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079083.1 NP_001072551.1 8 - 9 699 75
XM_012957278.2 XP_012812732.1 7 - 8 792 75
XM_018091349.1 XP_017946838.1 6 - 7 523 75
GenBank mRNA BC122064.1 - 8 - 9 699 75

View Full Product Details