Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lmnb3
Entrez Gene ID
733537
Gene Name
lamin B3
Gene Aliases
LB3, lamin-b3
Chromosome Location
Chr.3: 95699929 - 95717744 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083354.2 NP_001076823.1 9 - 10 1665 114
XM_012959159.2 XP_012814613.1 9 - 10 2194 114
GenBank mRNA BC135750.1 - 9 - 10 1665 114

View Full Product Details