Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ankrd2
Entrez Gene ID
100037897
Gene Name
ankyrin repeat domain 2 (stretch responsive muscle)
Gene Aliases
ankrd2-a, ankrd2-b, arpp
Chromosome Location
Chr.7: 24779865 - 24791243 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097332.1 NP_001090801.1 3 - 4 489 84
XM_012965830.2 XP_012821284.1 3 - 4 517 84
GenBank mRNA BC135222.1 - 3 - 4 489 84
CR434935.1 - 3 - 4 471 84

View Full Product Details