Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mip
Entrez Gene ID
100037914
Gene Name
major intrinsic protein of lens fiber
Gene Aliases
aqp0, lim1, mip26, mp26
Chromosome Location
Chr.2: 135268907 - 135274586 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097347.1 NP_001090816.1 1 - 2 567 92
GenBank mRNA BC135267.1 - 1 - 2 567 92

View Full Product Details