Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
shox2
Entrez Gene ID
100101703
Gene Name
short stature homeobox 2
Gene Aliases
og12, og12x, ogi2x, shot
Chromosome Location
Chr.5: 104063918 - 104073554 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100224.1 NP_001093694.1 3 - 4 709 85
XM_012962582.2 XP_012818036.1 3 - 4 949 85
GenBank mRNA DQ675518.1 - 3 - 4 709 85

View Full Product Details