Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kyat3
Entrez Gene ID
100124821
Gene Name
kynurenine aminotransferase 3
Gene Aliases
ccbl2
Chromosome Location
Chr.4: 95551272 - 95574839 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102788.1 NP_001096258.1 7 - 8 1645 96
GenBank mRNA BC084158.1 - 7 - 8 724 96
BC135439.1 - 7 - 8 1645 96

View Full Product Details