Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tsen15
Entrez Gene ID
100124883
Gene Name
tRNA splicing endonuclease subunit 15
Gene Aliases
-
Chromosome Location
Chr.4: 109615026 - 109631624 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102835.1 NP_001096305.2 2 - 3 204 103
XM_012960462.2 XP_012815916.1 2 - 3 451 103
GenBank mRNA BC135656.1 - 2 - 3 204 103

View Full Product Details