Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trmt44
Entrez Gene ID
100124986
Gene Name
tRNA methyltransferase 44 homolog
Gene Aliases
c4orf23, mettl19
Chromosome Location
Chr.1: 20770407 - 20793705 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012955724.2 XP_012811178.1 10 - 11 1875 78
XM_012955725.2 XP_012811179.1 10 - 11 1905 78
GenBank mRNA BC135974.1 - 10 - 11 1930 78

View Full Product Details