Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nmrk1
Entrez Gene ID
100125192
Gene Name
nicotinamide riboside kinase 1
Gene Aliases
c9orf95
Chromosome Location
Chr.1: 119245141 - 119260955 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103077.1 NP_001096547.1 7 - 8 927 103
XM_018094682.1 XP_017950171.1 6 - 7 861 103
XM_018094742.1 XP_017950231.1 6 - 7 859 103
GenBank mRNA BC136181.1 - 7 - 8 927 103

View Full Product Details