Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cmtm3
Entrez Gene ID
100127625
Gene Name
CKLF-like MARVEL transmembrane domain containing 3
Gene Aliases
-
Chromosome Location
Chr.4: 60431847 - 60443313 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112977.1 NP_001106448.1 3 - 4 395 125
GenBank mRNA BC153740.1 - 3 - 4 395 125

View Full Product Details