Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp2u1
Entrez Gene ID
100127656
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
-
Chromosome Location
Chr.1: 57521637 - 57536486 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113000.1 NP_001106471.1 2 - 3 1269 99
GenBank mRNA BC154092.1 - 2 - 3 1269 99

View Full Product Details