Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frs3
Entrez Gene ID
100101737
Gene Name
fibroblast growth factor receptor substrate 3
Gene Aliases
frs2b, frs2beta, snt-2, snt2
Chromosome Location
Chr.2: 62516963 - 62538622 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113669.1 NP_001107141.1 3 - 4 298 83
XM_012960231.2 XP_012815685.1 3 - 4 439 83
XM_012960256.2 XP_012815710.1 3 - 4 411 83
GenBank mRNA BC157660.1 - 3 - 4 298 83
BC171317.1 - 2 - 3 152 83
BC171319.1 - 2 - 3 152 83

View Full Product Details