Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
rabl6
Entrez Gene ID
100135091
Gene Name
RAB, member RAS oncogene family like 6
Gene Aliases
c9orf86
Chromosome Location
Chr.Un|NW_016683877: 2792 - 17015 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113830.1 NP_001107302.1 - 1847 99
GenBank mRNA BC155697.1 - - 1847 99
CR942773.1 - - 1584 99
CU464102.1 - - 1846 99

View Full Product Details