Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nom1
Entrez Gene ID
100135096
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
sgd1
Chromosome Location
Chr.6: 9629342 - 9654325 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113835.1 NP_001107307.1 11 - 12 1562 94
XM_012964243.2 XP_012819697.1 9 - 10 2472 94
GenBank mRNA BC155702.1 - 11 - 12 1562 94

View Full Product Details