Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nom1
Entrez Gene ID
100135096
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
sgd1
Chromosome Location
Chr.6: 9629342 - 9654325 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113835.1 NP_001107307.1 8 - 9 1169 75
GenBank mRNA BC155702.1 - 8 - 9 1169 75

View Full Product Details