Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sgo1
Entrez Gene ID
100135357
Gene Name
shugoshin 1
Gene Aliases
sgol1
Chromosome Location
Chr.6: 78483544 - 78492522 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114031.1 NP_001107503.1 3 - 4 392 69
XM_012964257.2 XP_012819711.1 - 443 69
GenBank mRNA BC157661.1 - 3 - 4 392 69
CR926350.2 - - 351 69

View Full Product Details