Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl47
Entrez Gene ID
100494080
Gene Name
mitochondrial ribosomal protein L47
Gene Aliases
-
Chromosome Location
Chr.5: 95112375 - 95118395 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114063.2 NP_001107535.2 5 - 6 579 90
GenBank mRNA CN108185.1 - 5 - 6 580 90
CX742176.1 - 5 - 6 507 90
EL854956.1 - 4 - 5 358 90

View Full Product Details