Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
apoo
Entrez Gene ID
100125130
Gene Name
apolipoprotein O
Gene Aliases
fam121b, my025
Chromosome Location
Chr.2: 38791831 - 38817822 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114243.1 NP_001107715.1 4 - 5 297 88
XM_012956316.1 XP_012811770.1 4 - 5 314 88
XM_012956317.2 XP_012811771.1 4 - 5 352 88
GenBank mRNA BC135301.1 - 3 - 4 282 88
BC155970.1 - 4 - 5 297 88

View Full Product Details