Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cbx3
Entrez Gene ID
100038121
Gene Name
chromobox 3
Gene Aliases
cbx3-a, cbx3-b, hech, hp1-gamma, hp1hs-gamma, xhp1g
Chromosome Location
Chr.6: 33427829 - 33439403 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122792.1 NP_001116264.1 4 - 5 455 109
XM_012964145.1 XP_012819599.1 4 - 5 738 109
XM_012964146.2 XP_012819600.1 4 - 5 695 109
XM_012964148.1 XP_012819602.1 3 - 4 625 109
XM_018094501.1 XP_017949990.1 4 - 5 485 109
XM_018094502.1 XP_017949991.1 5 - 6 861 109
XM_018094503.1 XP_017949992.1 3 - 4 323 109
GenBank mRNA BC161202.1 - 4 - 5 455 109
CT025206.2 - 2 - 3 305 109

View Full Product Details