Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
best4
Entrez Gene ID
100144681
Gene Name
bestrophin 4
Gene Aliases
vmd2l2
Chromosome Location
Chr.4: 72625093 - 72640179 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123442.1 NP_001116914.1 4 - 5 738 101
GenBank mRNA BC161756.1 - 4 - 5 738 101

View Full Product Details